Skip to Content
MilliporeSigma

P3C030D00

Pellicon® 3 Cassette with Ultracel® Membrane

filtration area 88 cm2, NMWCO 30 kDa, holdup volume, 3.6 mL (in feed channel)

Synonym(s):

Pellicon® 3 Cassette with Ultracel® 30 kDa Membrane, D screen, 88 cm2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41104903
NACRES:
NB.24
eCl@ss:
32031602
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

TPE seal, composite regenerated cellulose (CRC) membrane, polypropylene , polypropylene cap (for end), polypropylene screen

feature

holdup volume 3.6 mL (in feed channel)

manufacturer/tradename

Pellicon®

parameter

2.75 bar max. inlet pressure (40 psi) at 50 °C, 30 psi max. reverse press. exposure at 25 °C (for 24 hours), 4-6 L/min-m2 recirculation rate, 40 psi max. transmembrane pressure at 4-50 °C, 5.5 bar max. inlet pressure (80 psi) at 40 °C, 5.5 bar max. transmembrane pressure (80 psig) at 30 °C, 6.9 bar max. inlet pressure (100 psi), 80 psi max. transmembrane pressure at 4-40 °C

technique(s)

analytical sample preparation: suitable, buffer exchange: suitable, protein purification: suitable (concentration)

H

0.8 cm (0.33 in.)

L

20.6 cm (8.1 in.)

W

5.6 cm (2.2 in.)

filtration area

88 cm2

membrane area

88 cm2

gravimetric extractables

≤1500 mg/m2

matrix

Ultracel® (Membrane)

pore size

30 kDa NMWCO

pH

2-13

shipped in

ambient

General description

Pellicon® 3 filtration devices are advanced, high-performance cassettes ideal for today’s higher titer therapeutic antibodies as well as the more demanding filtration processes that require higher operating pressures, temperatures and caustic cleaning regimes.

The Pellicon® 3 Cassette with Ultracel® Membrane is designed for the filtration of therapeutic proteins (such as monoclonal antibodies) and recombinant and non-recombinant proteins.

This P3C030D00 Pellicon® 3 ultrafiltration device features a pore size of 30 kDa NMWCO, a filtration area of 88 cm2, a PLCTK membrane, and dimensions measuring 3.8 cm (H) x 17.8 cm (W) x 20.6 cm (L).

Features and Benefits

  • Rugged & reliable – ideally suited for today′s more demanding filtration processes
  • High performance – produces excellent resolution, yields and back-pressure resistance
  • Optimum product recovery – thanks to proven membrane technology
  • Void-free technology – leading-edge membranes resolve virtually any separation challenge
  • Defect-free, superior adhesion – Ultracel®uses composite membranes cast on a microporous substrate
  • Automated manufacturing – delivers unbeatable performance, consistency and reliability
  • Easy installation & care – simple to install and maintain

Analysis Note

Gravimetric Extractables: after 24 hours in water at controlled room temperature

Other Notes

These products are manufactured in a facility which adheres to Good Manufacturing Practices.

Legal Information

Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


Storage Class

10 - Combustible liquids

wgk

WGK 2



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library