Key Documents
SYNA751922
Synthware 90 degree vacuum adapter with glass stopcock
male joint: ST/NS 19/22
Synonym(s):
Synthware A751922, flow control adapter
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Joint
male joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A751922
General description
Used to regulate the flow of gases or liquids into reaction systems. Stopcock has a bore of 2 mm.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)