Key Documents
SYNA771410
Synthware inlet/thermometer adapter
joint: ST/NS 14/10
Synonym(s):
Synthware A771410, thermometer inlet adapter
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Joint
joint: ST/NS14/10
packaging
pack of 1 ea
manufacturer/tradename
A771410
Related Categories
General description
Synthware inlet or thermometer adapter with a compression cap and a Viton® O-ring to form a tight seal. Cap accommodates a 5.0-7.5mm O.D. plain stem thermometer. Supplied with one Nylon cap and one Viton® O-ring. For microscale applications. To use as syringe port, replace O-ring with PTFE-lined septum.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)