Skip to Content
MilliporeSigma
All Photos(1)

Documents

SYNA451922

Synthware inlet/thermometer adapter with one holed rubber adapter

joint: ST/NS 19/22

Synonym(s):

Synthware A451922, thermometer inlet adapter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121815
NACRES:
NB.51

Joint

joint: ST/NS19/22

packaging

pkg of 1 ea

manufacturer/tradename

Synthware A451922

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For use with gas inlet tubes or plain stem thermometers. Supplied with one holed rubber adapter.

Legal Information

Synthware is a trademark of Kemtech America Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service