Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

QBD11370

Sigma-Aldrich

DBCO-dPEG ®24-TFP ester

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C77H118F4N2O28
Molecular Weight:
1595.75
UNSPSC Code:
51171641
NACRES:
NA.22

Assay

>90%

form

solid or viscous liquid

reaction suitability

reaction type: click chemistry
reagent type: cross-linking reagent
reaction type: click chemistry

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(CCCC(NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCC(OC1=C(F)C(F)=CC(F)=C1F)=O)=O)N2CC3=CC=CC=C3C#CC4=C2C=CC=C4

InChI

1S/C77H118F4N2O28/c78-69-64-70(79)76(81)77(75(69)80)111-74(86)14-16-87-18-20-89-22-24-91-26-28-93-30-32-95-34-36-97-38-40-99-42-44-101-46-48-103-50-52-105-54-56-107-58-60-109-62-63-110-61-59-108-57-55-106-53-51-104-49-47-102-45-43-100-41-39-98-37-35-96-33

InChI key

DMZOSVFUYMWYBS-UHFFFAOYSA-N

Features and Benefits

DBCO-dPEG®₂₄-TFP ester contains a DBCO which can react with an azide through a copper-free "click" chemistry reaction and a TFP ester which can react with a primary amine.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service