Skip to Content
MilliporeSigma
All Photos(1)

Documents

QBD11167

Sigma-Aldrich

DOTA-tris(acid)-amido-dPEG ®11-Maleimide

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H85F6N7O25
Molecular Weight:
1310.24
UNSPSC Code:
12352106
NACRES:
NA.22

Assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: chelator
reagent type: cross-linking reagent

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

Features and Benefits

DOTA-tris(acid)-amido-dPEG ®₁₁-Maleimide contains a DOTA-tris(acid) metal chelator for radio-labelling and a thiol-reactive maleimide connected by a hydrophilic dPEG spacer.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service