Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

QBD10909

Sigma-Aldrich

m-dPEG®37-acid

>95% (HPLC)

Synonym(s):

m-PEG37-acid, Methyl-PEG37-acid, mPEG-acid, mPEG1500-acid, mPEG37-COOH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C76H152O39
Molecular Weight:
1690.00
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

Assay

>95% (HPLC)

form

solid or viscous liquid

reaction suitability

reaction type: Pegylations

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

Features and Benefits

m-dPEG37-acid is a long (112 atoms, 133.9 Å), methyl-capped, discrete chain length polyethylene glycol (dPEG) spacer. The reactive end of the molecule terminates in a propionic acid group. m-dPEG37-acid reacts with amine-functionalized surfaces (carbon nanotubes, nanoparticles made of silica or various metals, quantum dots, and similar types of products) or free amines on biomolecules. m-dPEG37-acid is a monodisperse equivalent of the polymeric PEG1500. When used to coat surfaces or modify biomolecules, m-dPEG37-acid reduces or eliminates non-specific binding and increases water solubility.

Legal Information

Products Protected under U.S. Patent #s 7,888,536 & 8,637,711 and European Patent #s 1,594,440 & 2,750,681
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service