Key Documents
PL800GH-8
Plas-Labs Hypalon glove box gloves
for 1 piece glove set, size 8 (for 8 in. ports)
Synonym(s):
Plas-Labs Glove Box
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
Pricing and availability is not currently available.
material
Hypalon®
size
8 (for 8 in. ports)
feature
reusable
packaging
pack of 1 pair
manufacturer/tradename
Plas-Labs 800GH-8
General description
Plas-Labs white Hypalon® rubber for resistance to abrasion, oxidizing chemicals, ozone and UV light
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
Legal Information
HaackerCalc is a trademark of ChemSW, Inc.
Hypalon is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)