Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

PL800GH-7

Plas-Labs Hypalon glove box gloves

size 7 (for 8 in. ports)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
46181504
NACRES:
NB.35
Pricing and availability is not currently available.

material

Hypalon®

size

7 (for 8 in. ports)

description

Includes hypalon sleeve and glove-over sleeve connector
Sold as a pair.

for 2 pieces glove set

feature

reusable

packaging

pack of 1 pair

manufacturer/tradename

Plas-Labs 800GH-7

Looking for similar products? Visit Product Comparison Guide

General description

Plas-Labs white Hypalon® rubber for resistance to abrasion, oxidizing chemicals, ozone and UV light

Note: Order sleeves, gloves, and glove over sleeve connectors separately.

Legal Information

HaackerCalc is a trademark of ChemSW, Inc.
Hypalon is a registered trademark of E. I. du Pont de Nemours and Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Guten Tag, können Sie mir die Materialstärke nennen?

    1 answer
    1. This product is comprised of 2 pieces. The thickness of the sleeve is 15 mm and the thickness of the glove is 13 mm.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service