Skip to Content
MilliporeSigma
All Photos(3)

Documents

P66203

Sigma-Aldrich

trans-3-(3-Pyridyl)acrylic acid

99%

Synonym(s):

3-Pyridineacrylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7NO2
CAS Number:
Molecular Weight:
149.15
Beilstein:
113290
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

mp

232-235 °C (dec.) (lit.)

SMILES string

[H]\C(=C(\[H])c1cccnc1)C(O)=O

InChI

1S/C8H7NO2/c10-8(11)4-3-7-2-1-5-9-6-7/h1-6H,(H,10,11)/b4-3+

InChI key

VUVORVXMOLQFMO-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

trans-3-(3-Pyridyl)acrylic acid, a bifunctional ligand with both hard and soft coordination sites, is mainly used in constructing heterometallic complexes.

Application

trans-3-(3-Pyridyl)acrylic acid has been used as an organic linker to connect d and f block metals to form a 3D heterometallic coordination polymer with luminescence property. It may also be used in the synthesis of ethyl trans-3-(3-pyridyl)acrylate via esterification.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Uranyl Ion Complexes with trans?3?(3?Pyridyl) acrylic Acid Including a Uranyl?Copper (II) Heterometallic Framework.
Thuery P
European Journal of Inorganic Chemistry, 2014(28), 4772-4778 (2014)
Novel coordination polymers and structural systematics in the hydrothermal M, M? trans-3 (-3-pyridyl) acrylic acid system.
Gunning N
Dalton Transactions, 16, 2788-2792 (2005)
Matrix?assisted laser desorption mass spectrometry of homopolymer oligodeoxyribonucleotides. Influence of base composition on the mass spectrometric response.
Schneider K
Journal of Mass Spectrometry : Jms, 28(11), 1353-1361 (1993)
Interparticle spacing control in the superlattices of carboxylic acid-capped gold nanoparticles by hydrogen-bonding mediation.
Yao H
Langmuir, 20(23), 10317-10323 (2004)
Combination of covalent and hydrogen bonding in the formation of 3D uranyl-carboxylate networks.
Dalai S
Inorgorganica Chimica Acta, 363(13), 3407-3412 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service