Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.07468

Sigma-Aldrich

3-Pyridinecarbaldehyde

for synthesis

Synonym(s):

3-Pyridinecarbaldehyde, Nicotinaldehyde, Pyridine-3-aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
3-(CHO)C5H4N
CAS Number:
Molecular Weight:
107.11
MDL number:
UNSPSC Code:
12352114
EC Index Number:
207-900-4
NACRES:
NA.22

vapor pressure

0.3 hPa ( 20 °C)

Quality Level

Assay

≥97.0% (GC)

form

liquid

potency

2355 mg/kg LD50, oral (Rat)

pH

5.4 (20 °C, 111 g/L in H2O)

mp

7 °C

transition temp

flash point 84 °C

density

1.14 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

n1cc(ccc1)C=O

InChI

1S/C6H5NO/c8-5-6-2-1-3-7-4-6/h1-5H

InChI key

QJZUKDFHGGYHMC-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.141 - 1.145
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service