Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P27127

Sigma-Aldrich

Phenylmercuric acetate

97%

Synonym(s):

Phenylmercury acetate, Mercury phenyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5HgOCOCH3
CAS Number:
Molecular Weight:
336.74
Beilstein:
3662930
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

148-151 °C (lit.)

SMILES string

CC(=O)O[Hg]c1ccccc1

InChI

1S/C6H5.C2H4O2.Hg/c1-2-4-6-5-3-1;1-2(3)4;/h1-5H;1H3,(H,3,4);/q;;+1/p-1

InChI key

XEBWQGVWTUSTLN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Phenylmercuric acetate (PMA) can react with a palladium salt such as palladium acetate to form an arylpalladium salt in situ, which then reacts with primary or secondary allylic alcohols to produce 3-aryl aldehydes. It can also be used in the multi-step synthesis of 2,3,4,6-tetra-O-acetyl-α-D-glucopyranosyl thiol via Floyd′s pathway.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT RE 1

Target Organs

Kidney,Central nervous system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of aromatic and indole alpha-glucosinolates.
Vo Quan V, et al.
Carbohydrate Research, 455, 45-53 (2018)
The arylation of allylic alcohols with organopalladium compounds. A new synthesis of 3-aryl aldehydes and ketones.
Heck R F, et al.
Journal of the American Chemical Society, 90(20), 5526-5531 (1968)
S K Ghosh et al.
Journal of applied microbiology, 102(2), 375-383 (2007-01-24)
Mercury compounds are highly toxic to all types of living cells. Isolated yeast strains of Rhodotorula rubra showed high and low resistance pattern towards mercury and organomercurial compounds. To investigate the basis of differential sensitivity of these two types of
Harry J Beaulieu et al.
Journal of occupational and environmental hygiene, 5(6), 360-366 (2008-03-28)
Phenyl mercuric acetate (PMA) historically has been used as a catalyst in polyurethane systems. In the 1950s-1970s, PMA was used as a catalyst in the 3M Tartan brand polyurethane flexible floors that were installed commonly in school gymnasiums. Mercury vapor
Lilla Landeck et al.
International journal of dermatology, 50(7), 806-810 (2011-06-28)
Some authors have claimed a decreased cell-mediated immunity among atopic individuals, which would lead to observations of decreased rates of allergic contact dermatitis (ACD). The purpose of our study was to investigate contact sensitization patterns in atopic subjects compared with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service