Skip to Content
MilliporeSigma
All Photos(1)

Documents

P23903

Sigma-Aldrich

1,4-Phenylenediacrylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH=CHCO2H)2
CAS Number:
Molecular Weight:
218.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

mp

>300 °C (lit.)

SMILES string

OC(=O)\C=C\c1ccc(\C=C\C(O)=O)cc1

InChI

1S/C12H10O4/c13-11(14)7-5-9-1-2-10(4-3-9)6-8-12(15)16/h1-8H,(H,13,14)(H,15,16)/b7-5+,8-6+

InChI key

AAFXQFIGKBLKMC-KQQUZDAGSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jack M Woolley et al.
Physical chemistry chemical physics : PCCP, 21(26), 14350-14356 (2019-02-01)
Photoisomerisation has been shown to be an efficient excited-state relaxation mechanism for a variety of nature-based and artificial-based molecular systems. Here we report on the excited-state relaxation dynamics and consequent photostability of a symmetrically functionalised cinnamate by transient electronic absorption

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service