Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

225266

Sigma-Aldrich

Biphenyl-4,4′-dicarboxylic acid

97%

Synonym(s):

4-(4-propan-2-yloxycarbonylphenyl)benzoic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CC6H4C6H4CO2H
CAS Number:
Molecular Weight:
242.23
Beilstein:
523308
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

97%

mp

>300 °C (lit.)

SMILES string

OC(=O)c1ccc(cc1)-c2ccc(cc2)C(O)=O

InChI

1S/C14H10O4/c15-13(16)11-5-1-9(2-6-11)10-3-7-12(8-4-10)14(17)18/h1-8H,(H,15,16)(H,17,18)

InChI key

NEQFBGHQPUXOFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Biphenyl-4,4′-dicarboxylic acid (BPDC) belongs to the class of monomers known as aromatic dicarboxylic acids. It is used to prepare BPDC-based polymers, which exhibit excellent thermal stability, making them suitable for applications where high-temperature resistance is required. These polymers also demonstrate high electrical insulation properties due to their low dielectric constant and high breakdown strength. They find applications in the electrical and electronics industry for insulation, encapsulation, and fabrication of high-performance electronic devices.[1]

Application

Biphenyl-4,4′-dicarboxylic acid can be used:
  • As a key component In the synthesis of nematic aromatic polyesters which are valuable in the development of liquid crystal polymers with potential applications in electronics, optical devices automotive, and textiles.
  • As a linker or ligand in the synthesis of metal-organic frameworks (MOFs) for a wide range of potential applications in areas such as gas storage, catalysis, sensing, and separation technology.
  • As a building block in biomedical applications for the synthesis of various pharmaceutical compounds and materials with biological activity. It can also be utilized in the development of drug delivery systems and biomaterials due to its chemical properties.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eugene R Zubarev et al.
Organic letters, 8(7), 1367-1370 (2006-03-28)
[reaction: see text] We describe a synthesis of a dendrimer-like amphiphile containing a flat rigid core and 12 hydrophobic and hydrophilic arms. We employ a modular approach based on stepwise protection chemistry starting from simple building blocks. The key feature

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service