Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P22303

Sigma-Aldrich

1-Phenyl-1-cyclohexene

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H9
CAS Number:
Molecular Weight:
158.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

refractive index

n20/D 1.57 (lit.)

bp

251-253 °C (lit.)

mp

−11 °C (lit.)

density

0.994 g/mL at 25 °C (lit.)

SMILES string

C1CCC(=CC1)c2ccccc2

InChI

1S/C12H14/c1-3-7-11(8-4-1)12-9-5-2-6-10-12/h1,3-4,7-9H,2,5-6,10H2

InChI key

WCMSFBRREKZZFL-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D H Young et al.
Bioorganic & medicinal chemistry letters, 11(11), 1393-1396 (2001-05-30)
Phenylcyclohexenes (PCHs) [e.g., trans-4-nitro-5-(2,3,4-trimethoxyphenyl)cyclohexene, 2d] were found to bind weakly to the colchicine site of bovine tubulin, but are the first mimics of colchicine found to have high activity towards plant cells. Structure-activity relationships for PCHs and biphenyl AC-ring analogues
B R Martin et al.
Drug metabolism and disposition: the biological fate of chemicals, 10(6), 685-689 (1982-11-01)
The in vitro metabolism of 1-3H-phenyl-1-cyclohexene (3H-PC) was studied in a crude microsomal preparation from mouse livers. The major routes of metabolism were allylic hydroxylation, oxidation of the allylic alcohol, and epoxidation-hydrolysis. The following metabolites were identified by comparison with
S Chakrabarti et al.
Toxicology and applied pharmacology, 69(2), 179-184 (1983-06-30)
The metabolic disposition of 1-[14C]phenylcyclohexene ([14C]PC) was examined in rats after ip or iv drug administration. Radioactivity, which was accumulated by various organs, peaked within 30 min after ip administration of [14C]PC (0.21 mg/kg). A significant amount of this radioactivity
A S Freeman et al.
Journal of pharmaceutical sciences, 70(9), 1002-1004 (1981-09-01)
Parsley cigarettes containing [3H]phencyclidine were machine smoked, and the mainstream smoke was trapped in glass wool filters. Radioactivity was extracted from these filters with chloroform. The average recoveries of radioactivity were 76, 85, 70, and 69% for cigarettes containing 3
R L Beard et al.
Bioorganic & medicinal chemistry letters, 11(6), 765-768 (2001-03-30)
Retinoids are natural and synthetic analogues of the hormone retinoic acid. Systemic retinoid agonist therapy is usually associated with toxic side effects, such as mucocutaneous toxicity, which may be alleviated by the use of topical retinoid antagonists. We report the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service