Merck
  • Phenylcyclohexene and phenylcyclohexadiene substituted compounds having retinoid antagonist activity.
Phenylcyclohexene and phenylcyclohexadiene substituted compounds having retinoid antagonist activity.
Bioorganic & medicinal chemistry letters (2001-03-30)
R L Beard, E S Klein, A M Standeven, M Escobar, R A Chandraratna
ABSTRACT
Retinoids are natural and synthetic analogues of the hormone retinoic acid. Systemic retinoid agonist therapy is usually associated with toxic side effects, such as mucocutaneous toxicity, which may be alleviated by the use of topical retinoid antagonists. We report the synthesis and biological activity of a new series of potent, RAR-specific antagonists substituted with phenylcyclohexene and phenylcyclohexadiene groups.
Sigma-Aldrich
1-Phenyl-1-cyclohexene, 95%