Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

N9829

Sigma-Aldrich

3-Nitroaniline

98%

Synonym(s):

1-Amino-3-nitrobenzene, 3-Aminonitrobenzene, 3-Nitrobenzenamine, 3-Nitrophenylamine, m-Aminonitrobenze, m-Aminonitrobenzene, m-Nitroaminobenzene, m-Nitroaniline, m-Nitrophenylamine, meta-Nitroaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4NH2
CAS Number:
Molecular Weight:
138.12
Beilstein:
636962
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1 mmHg ( 119 °C)

Assay

98%

form

fibers
powder, crystals or chunks

mp

111-114 °C (lit.)

SMILES string

Nc1cccc(c1)[N+]([O-])=O

InChI

1S/C6H6N2O2/c7-5-2-1-3-6(4-5)8(9)10/h1-4H,7H2

InChI key

XJCVRTZCHMZPBD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - STOT RE 2 Oral

Target Organs

Kidney,Liver,Heart,Blood

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

384.8 °F - closed cup

Flash Point(C)

196.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yulia S Sotnikova et al.
Talanta, 220, 121400-121400 (2020-09-16)
The properties of chromatographic columns are largely determined by functional groups located on the sorbent surface. For monolithic columns, surface functional groups can be created during synthesis stage or by chemical bonding with the complete surface of the sorbent. One
Jonas Winters et al.
Physical chemistry chemical physics : PCCP, 21(7), 4053-4062 (2019-02-05)
Polyaramids are a class of high-performance polymers, known for their high mechanical strength and chemical and thermal stability. Their ability to create a network of intermolecular hydrogen bonds causes them to be very poorly soluble in conventional solvents. Hazardous solvents
L A Donaldson et al.
Journal of microscopy, 259(3), 228-236 (2015-05-01)
A novel approach to nanoscale detection of cell wall porosity using confocal fluorescence microscopy is described. Infiltration of cell walls with a range of nitrophenyl-substituted carbohydrates of different molecular weights was assessed by measuring changes in the intensity of lignin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service