Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

616508

Sigma-Aldrich

3-Nitroaniline-2,4,5,6-d4

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D4H2N2O2
Molecular Weight:
142.15
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

99% (CP)

form

solid

mp

112-114 °C (lit.)

mass shift

M+4

storage temp.

2-8°C

SMILES string

[2H]c1c([2H])c(N)c([2H])c(c1[2H])[N+]([O-])=O

InChI

1S/C6H6N2O2/c7-5-2-1-3-6(4-5)8(9)10/h1-4H,7H2/i1D,2D,3D,4D

InChI key

XJCVRTZCHMZPBD-RHQRLBAQSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

384.8 °F - closed cup

Flash Point(C)

196.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service