Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M38214

Sigma-Aldrich

1-Methylcyclohexanol

96%

Synonym(s):

1-Hydroxy-1-methylcyclohexane, 1-Methyl-1-cyclohexanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H10OH
CAS Number:
Molecular Weight:
114.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.4585 (lit.)

bp

168 °C/752 mmHg (lit.)

mp

24-26 °C (lit.)

density

0.919 g/mL at 25 °C (lit.)

SMILES string

CC1(O)CCCCC1

InChI

1S/C7H14O/c1-7(8)5-3-2-4-6-7/h8H,2-6H2,1H3

InChI key

VTBOTOBFGSVRMA-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Kellett et al.
Journal of computer-aided molecular design, 35(1), 95-104 (2020-10-11)
We investigate the binding of native β-cyclodextrin (β-CD) and eight novel β-CD derivatives with two different guest compounds, using isothermal calorimetry and 2D NOESY NMR. In all cases, the stoichiometry is 1:1 and binding is exothermic. Overall, modifications at the
Liping Kong et al.
Bioresource technology, 299, 122582-122582 (2019-12-27)
Direct hydrogenolysis of Kraft lignin was catalyzed over a series of supported Ni or Re catalysts in ethanol solvent. The best results showed that the oil yield of 96.70 wt% was obtained with less char formation at 330 °C for 3 h over
Verónica Cattaneo et al.
Neurobiology of learning and memory, 172, 107233-107233 (2020-05-04)
Intellectual disability is a common feature in genetic disorders with enhanced RAS-ERK1/2 signaling, including neurofibromatosis type 1 (NF1) and Noonan syndrome (NS). Additional training trials and additional spacing between trials, respectively, restores memory deficits in animal models of NF1 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service