Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

178829

Sigma-Aldrich

trans-2-Methylcyclohexanol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H10OH
CAS Number:
Molecular Weight:
114.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.461 (lit.)

bp

167.2-167.6 °C (lit.)

mp

−21 °C (lit.)

density

0.924 g/mL at 25 °C (lit.)

SMILES string

C[C@@H]1CCCC[C@H]1O

InChI

1S/C7H14O/c1-6-4-2-3-5-7(6)8/h6-8H,2-5H2,1H3/t6-,7-/m1/s1

InChI key

NDVWOBYBJYUSMF-RNFRBKRXSA-N

General description

2-Methylcyclohexanol binds to the exo-site of the small molecules.

Application

2-Methylcyclohexanol was used in the fragment-based crystallographic screening against HIV protease.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander L Perryman et al.
Chemical biology & drug design, 75(3), 257-268 (2010-07-28)
We have employed a fragment-based screen against wild-type (NL4-3) HIV protease (PR) using the Active Sight fragment library and X-ray crystallography. The experiments reveal two new binding sites for small molecules. PR was co-crystallized with fragments, or crystals were soaked

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service