Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

M29304

Sigma-Aldrich

N-Methylaniline

98%

Synonym(s):

Monomethylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHCH3
CAS Number:
Molecular Weight:
107.15
Beilstein:
741982
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.571 (lit.)

bp

196 °C (lit.)

mp

−57 °C (lit.)

density

0.989 g/mL at 25 °C (lit.)

SMILES string

CNc1ccccc1

InChI

1S/C7H9N/c1-8-7-5-3-2-4-6-7/h2-6,8H,1H3

InChI key

AFBPFSWMIHJQDM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Methylaniline is a general reagent used:
  • In the preparation of phosphoramidite ligands for the rhodium-catalyzed asymmetric hydroboration.[1]
  • As a formylating reagent in the C3-selective formylation of indoles.[2]
  • In the synthesis of a rhodol-based fluorescent probe, HKGreen-3, for sensing peroxynitrite.[3]
  • As a key starting material in the synthesis of AKT (protein kinase B) inhibitor-IV.[4]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - STOT RE 2 Oral

Target Organs

Liver,spleen,Bone marrow

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and biological evaluation of analogues of AKT (protein kinase B) inhibitor-IV.
Sun Q, et al.
Journal of Medicinal Chemistry, 54(5), 1126-1139 (2011)
Efficient amide-directed catalytic asymmetric hydroboration.
Smith S M, et al.
Journal of the American Chemical Society, 130(12), 3734-3735 (2008)
HKGreen-3: a rhodol-based fluorescent probe for peroxynitrite.
Peng T and Yang D
Organic Letters, 12(21), 4932-4935 (2010)
V Arjunan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 72(2), 436-444 (2008-12-17)
The Fourier transform infrared (FTIR) and FT-Raman spectra of 2-chloro-4-methylaniline and 2-chloro-6-methylaniline have been measured in the range 4000-400 and 4000-100cm(-1), respectively. Utilising the observed FTIR and FT-Raman data, a complete vibrational assignment and analysis of the fundamental modes of
Dongmei Li et al.
Dalton transactions (Cambridge, England : 2003), (2)(2), 291-297 (2008-12-18)
The mechanism of N-dealkylation of N-cyclopropyl-N-methylaniline () catalyzed by cytochrome P450 (P450) was investigated using density functional theory. This reaction involves two steps. The first one is a Calpha-H hydroxylation on the N-substituent to form a carbinolaniline complex, and the

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service