Skip to Content
MilliporeSigma

8.06096

N-Methylaniline

for synthesis

Synonym(s):

N-Methylaniline, Monomethylaniline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C7H9N
CAS Number:
Molecular Weight:
107.15
UNSPSC Code:
12352116
EC Index Number:
202-870-9
NACRES:
NA.22
MDL number:
Assay:
≥98.0% (GC)
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

0.5 hPa ( 20 °C)

Quality Segment

assay

≥98.0% (GC)

form

liquid

autoignition temp.

500 °C

potency

360 mg/kg LD50, oral (Rat)

pH

7.6 (20 °C, 1 g/L in H2O)

bp

196 °C/1013 hPa

mp

-57 °C

transition temp

flash point 86 °C

density

0.99 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(C)c1ccccc1

InChI

1S/C7H9N/c1-8-7-5-3-2-4-6-7/h2-6,8H,1H3

InChI key

AFBPFSWMIHJQDM-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.985 - 0.986
Identity (IR): passes test


Still not finding the right product?

Explore all of our products under N-Methylaniline


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - STOT RE 2 Oral

Storage Class

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

185.0 °F - closed cup

flash_point_c

85 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library