Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

M27409

Sigma-Aldrich

3-Methyladipic acid

99%

Synonym(s):

3-Methylhexanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2CH2CH(CH3)CH2CO2H
CAS Number:
Molecular Weight:
160.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

bp

230 °C/30 mmHg (lit.)

mp

100-102 °C (lit.)

SMILES string

CC(CCC(O)=O)CC(O)=O

InChI

1S/C7H12O4/c1-5(4-7(10)11)2-3-6(8)9/h5H,2-4H2,1H3,(H,8,9)(H,10,11)

InChI key

SYEOWUNSTUDKGM-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joaquín Barberá et al.
Journal of the American Chemical Society, 127(1), 458-464 (2005-01-06)
The work described here concerns a challenge of general interest in supramolecular chemistry: the achievement of chiral helical organizations with controlled structures. This work provides a strategy to obtain supramolecular polymers in which a chiral helical conformation has been induced
3-Methyladipate excretion in animals fed a phytol supplement with reference to Refsum's disease.
S Krywawych et al.
Journal of inherited metabolic disease, 8 Suppl 2, 147-148 (1985-01-01)
G Steen et al.
Acta neurologica Scandinavica, 68(4), 231-240 (1983-10-01)
The urinary excretion of organic acids was examined in 509 cases with multiple sclerosis and in 50 age- and sex-matched controls. The concentrations of the acids were related to creatinine. No differences were found for compounds such as glycolic acid
3-Methyladipate excretion in Refsum's disease.
D P Brenton et al.
Lancet (London, England), 1(8272), 624-624 (1982-03-13)
L Williams-Seton et al.
Journal of pharmaceutical sciences, 89(3), 346-354 (2000-03-09)
The variation in physical properties of crystals grown in the presence of additives or impurities have previously been attributed to lattice disorder developed during crystallization. Adipic acid crystallized in the presence of a variety of stereochemically related impurities typifies such

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service