Documents
S5641
Sigma-Aldrich
Scienceware® Coplin staining jar with screw cap
high-density polypropylene
Synonym(s):
coplin jar, microscope slide staining jar
About This Item
UNSPSC Code:
41181600
NACRES:
MA.01
material
flat screw cap (53 mm in diameter)
high-density polypropylene
packaging
pack of 12 ea
manufacturer/tradename
Bel-Art F44208-1000
General description
The flat-topped polypropylene threaded closure is 53 mm in diameter, features a large milled edge for a convenient hand grip and has deep sealing screw threads. Five internal slots store up to 10 standard 1 x 3" or 25 x 75 mm slides back to back. The base diameter is 59.7 mm (23/8"), height is 114 mm (41/2"). Steam autoclavable at 121°C (250°F), with lid detached during autoclaving.
  • Durable polypropylene construction
  • Sealing ring makes jar watertight
  • Stores up to 10 standard slides
  • Autoclavable
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Visit the Document Library
Yan Wang et al.
Chemosphere, 267, 128870-128870 (2020-11-12)
The pathogenesis of pulmonary fibrosis diseases is considered to be related with environmental exposures, but the exact mechanism is unclear and there are no effective treatments. The contribution of epithelial-mesenchymal transition (EMT) to lung fibrosis has been controversial. It was
Articles
Available Fluorescent in situ hybridization (FISH) procedures, reagents and equipment.
Available Fluorescent in situ hybridization (FISH) procedures, reagents and equipment.
Available Fluorescent in situ hybridization (FISH) procedures, reagents and equipment.
Available Fluorescent in situ hybridization (FISH) procedures, reagents and equipment.
Protocols
Use this protocol to for the entire immunohistochemistry (IHC) procedure through staining and visualization of specific antigens in paraffin-embedded tissue sections.
Use this protocol to for the entire immunohistochemistry (IHC) procedure through staining and visualization of specific antigens in paraffin-embedded tissue sections.
Use this protocol to for the entire immunohistochemistry (IHC) procedure through staining and visualization of specific antigens in paraffin-embedded tissue sections.
Use this protocol to for the entire immunohistochemistry (IHC) procedure through staining and visualization of specific antigens in paraffin-embedded tissue sections.