Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M070000

Sigma-Aldrich

4,5-Dicyanoimidazole

Synonym(s):

4,5-Imidazoledicarbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H2N4
CAS Number:
Molecular Weight:
118.10
Beilstein:
118208
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

type

for DNA synthesis

product line

Proligo Reagents

Assay

≥99.0% (reversed phase HPLC)

impurities

≤300 ppm water content (Karl Fischer)

color

solid white

mp

168-175 °C (lit.)
174-176 °C

suitability

conforms to structure for H-NMR

SMILES string

N#Cc1nc[nH]c1C#N

InChI

1S/C5H2N4/c6-1-4-5(2-7)9-3-8-4/h3H,(H,8,9)

InChI key

XGDRLCRGKUCBQL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew T Holden et al.
Analytical chemistry, 87(22), 11420-11428 (2015-10-24)
The photolithographic fabrication of high-density DNA and RNA arrays on flexible and transparent plastic substrates is reported. The substrates are thin sheets of poly(ethylene terephthalate) (PET) coated with cross-linked polymer multilayers that present hydroxyl groups suitable for conventional phosphoramidite-based nucleic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service