Skip to Content
MilliporeSigma
All Photos(1)

Documents

L084000

Sigma-Aldrich

20% Diethylamine in Acetonitrile

configured for ÄKTA® andOligoPilot®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352116
NACRES:
NA.21

type

for DNA synthesis

Quality Level

product line

Proligo Reagents

impurities

≤250 ppm water content (Karl Fischer)

color

clear

compatibility

configured for ÄKTA® andOligoPilot®

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Diethylamine is a secondary aliphatic amine with both N-substituents as ethyl groups. It is a conjugate base of a diethylammonium.

Components

Diethylamine / Acetonitrile 20/80 v/v

Legal Information

OligoPilot is a registered trademark of Cytiva
ÄKTA is a registered trademark of Cytiva

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service