Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

JKA3106

Sigma-Aldrich

mPEG5K

average MN 5,000, Phosphate

Synonym(s):

Methoxy Polyethylene Glycol Phosphate, Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2CH2O)nCH2CH2OPO(OH)2
UNSPSC Code:
51171641
NACRES:
NA.23

Product Name

mPEG5K-Phosphate, average Mn 5000

form

solid

mol wt

average Mn 5000

storage temp.

−20°C

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service