Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I2800

Sigma-Aldrich

Indene

contains 50-100 ppm tert-butylcatechol as stabilizer, technical grade, ≥90%

Synonym(s):

Indonaphthene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8
CAS Number:
Molecular Weight:
116.16
Beilstein:
635873
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

≥90%

form

liquid

contains

50-100 ppm tert-butylcatechol as stabilizer

refractive index

n20/D 1.595 (lit.)

bp

181-182 °C (lit.)

mp

−5-−3 °C (lit.)

density

0.996 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C1C=Cc2ccccc12

InChI

1S/C9H8/c1-2-5-9-7-3-6-8(9)4-1/h1-6H,7H2

InChI key

YBYIRNPNPLQARY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucas J Gursky et al.
Applied microbiology and biotechnology, 85(4), 995-1004 (2009-07-02)
The styAB genes from Pseudomonas putida CA-3, which encode styrene monooxygenase, were subjected to three rounds of in vitro evolution using error-prone polymerase chain reaction with a view to improving the rate of styrene oxide and indene oxide formation. Improvements
Adam C Glass et al.
Organic letters, 10(21), 4855-4857 (2008-10-07)
A new methodology for the preparation of substituted naphthalenes starting from readily available indenones, organometal reagents, and trimethylsilyldiazomethane via a catalytic rearrangement process is described. Hindered biaryl naphthalenes, including triortho-substituted biaryls, can be accessed through our method. Our results are
Regioselective synthesis of indenols by rhodium-catalyzed C-H activation and carbocyclization of aryl ketones and alkynes.
Krishnamoorthy Muralirajan et al.
Angewandte Chemie (International ed. in English), 50(18), 4169-4172 (2011-03-31)
Two-step synthesis of stable dioxadicarbaporphyrins from bis(3-indenyl)methane.
Timothy D Lash et al.
Angewandte Chemie (International ed. in English), 51(43), 10871-10875 (2012-09-25)
Ermitas Alcalde et al.
Journal of medicinal chemistry, 52(3), 675-687 (2009-01-23)
Scaffold selection involving an indole-to-indene core change led to the discovery of a series of indenylsulfonamides that act as 5-HT6 serotonin receptor agonists. The variety of the targeted ligands and their synthetic complexity required multistep synthetic approaches. The novel indenylsulfonamides

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service