Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I10457

Sigma-Aldrich

3-Iodopropionic acid

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ICH2CH2CO2H
CAS Number:
Molecular Weight:
199.98
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

mp

80-83 °C (lit.)

SMILES string

OC(=O)CCI

InChI

1S/C3H5IO2/c4-2-1-3(5)6/h1-2H2,(H,5,6)

InChI key

KMRNTNDWADEIIX-UHFFFAOYSA-N

Other Notes

Remainder 3-chloropropionic acid

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raphael C L Machado et al.
Molecules (Basel, Switzerland), 24(23) (2019-11-27)
Compared to conventional spectroscopy or chromatography analysis, chemical sensing based on colorimetric changes offers an alternative to monitor potential metal hazards in aqueous environment through rapid and low-cost colorimetric changes which can be easily interpreted. In this work poly(ethylene glycol)
Eugénie Grigorian et al.
Frontiers in microbiology, 12, 725997-725997 (2021-10-09)
L-2-halocid dehalogenases (L-2-HADs) have been mainly characterized from terrestrial polluted environments. By contrast, knowledge is still scarce about their role in detoxification of predominant halocarbons in marine environments. Here, phylogenetic analyses showed a wide diversity of homologous L-2-HADs, especially among
Janina Kabatc et al.
Journal of pharmaceutical and biomedical analysis, 114, 433-440 (2015-06-30)
Non-covalent interaction of hemicyanine dyes, derivatives of 2,3,3-trimethyl-3H-indolium with bovine serum albumin has been studied by spectral method. For this purpose, three hemicyanine dyes containing N-(2-carboxyethyl)-2,3,3-trimethyl-3H-indolium moiety were synthesized and their UV/Vis and fluorescence spectra, aggregation, photostability and association with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service