Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

128163

Sigma-Aldrich

Ethyl 3-bromopropionate

98%

Synonym(s):

β-Bromopropionic acid ethyl ester, 2-Ethoxycarbonylethyl bromide, 3-Bromopropanoic acid ethyl ester, 3-Bromopropionic acid ethyl ester, Ethyl β-bromopropionate, Ethyl 3-bromopropanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CH2COOC2H5
CAS Number:
Molecular Weight:
181.03
Beilstein:
1700933
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

refractive index

n20/D 1.452 (lit.)

bp

135-136 °C/50 mmHg (lit.)

solubility

H2O: soluble
alcohol: soluble
benzene: soluble
chloroform: soluble
diethyl ether: soluble

density

1.412 g/mL at 25 °C (lit.)

functional group

bromo
ester

SMILES string

CCOC(=O)CCBr

InChI

1S/C5H9BrO2/c1-2-8-5(7)3-4-6/h2-4H2,1H3

InChI key

FQTIYMRSUOADDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethyl 3-bromopropionate was used in the preparation of (3′,4′-dimethoxyphenyl)-5-oxopentanoic acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel 2-amino-4-aryl-substituted-and 2-amino-4, 5-disubstituted-thiazoles.
Berlin KD and Herd MD.
Proceedings of the Oklahoma Academy of Science, 71, 29-33 (1991)
Chenxi Li et al.
Molecular pharmaceutics, 16(6), 2648-2660 (2019-05-03)
The complement system plays an important role in host innate immunity, and its activation can be exploited as a potential strategy for vaccine adjuvants. Herein, a pH-responsive micellar vaccine platform (COOH-NPs) was developed using a carboxyl-modified diblock copolymer of poly(2-ethyl-2-oxazoline)-poly(d,l-lactide)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service