Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF98899687

Praseodymium

foil, 50x50mm, thickness 0.25mm, as rolled, 99%

Synonym(s):

Praseodymium, PR000230

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pr
CAS Number:
Molecular Weight:
140.91
MDL number:
UNSPSC Code:
12141610
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

foil

manufacturer/tradename

Goodfellow 988-996-87

resistivity

68 μΩ-cm, 20°C

bp

3520 °C (lit.)

mp

931 °C (lit.)

density

6.71 g/mL at 25 °C (lit.)

SMILES string

[Pr]

InChI

1S/Pr

InChI key

PUDIUYLPXJFUGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohamadreza K Bakht et al.
Annals of nuclear medicine, 25(8), 529-535 (2011-07-02)
Radionuclides of rare earth elements are gaining importance as emerging therapeutic agents in nuclear medicine. β(-)-particle emitter 142Pr [T (1/2) = 19.12 h, E(-)β = 2.162 MeV (96.3%), Eγ = 1575 keV (3.7%)] is one of the praseodymium-141 (100% abundant)
Wei Gao et al.
Journal of nanoscience and nanotechnology, 14(6), 4308-4312 (2014-04-18)
Pr3+/Yb3+ co-doped hexagonal NaYF4(beta-NaYF4) microprisms were synthesized by the hydrothermal method, and ethylenediaminetetraacetic acid (EDTA) was introduced to control the size of the microcrystal samples. Bright upconverted fluorescence emission was observed when the samples were excited with an infrared (IR)
Sunbin Liu et al.
eLife, 4, e07320-e07320 (2015-07-15)
Prp3 is an essential U4/U6 di-snRNP-associated protein whose functions and molecular mechanisms in pre-mRNA splicing are presently poorly understood. We show by structural and biochemical analyses that Prp3 contains a bipartite U4/U6 di-snRNA-binding region comprising an expanded ferredoxin-like fold, which
Uwe Lindner et al.
Analytical and bioanalytical chemistry, 407(9), 2415-2422 (2014-12-10)
Hydrophilic interaction chromatography (HILIC) coupled with inductively coupled plasma mass spectrometry (ICP-MS) were optimised for the direct determination of gadolinium-based contrast agents in tap water. In comparison to our previous work, a new developed zwitterionic HILIC column (ZIC-cHILIC) was used
Carmen Cavallo et al.
Scientific reports, 5, 16785-16785 (2015-11-19)
Solid solutions of the rare earth (RE) cations Pr(3+), Nd(3+), Sm(3+), Gd(3+), Er(3+) and Yb(3+) in anatase TiO2 have been synthesized as mesoporous beads in the concentration range 0.1-0.3% of metal atoms. The solid solutions were have been characterized by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service