Skip to Content
MilliporeSigma
All Photos(2)

Documents

GF85844740

Zirconium

powder, 45 max. part. size (micron), weight 200 g, purity 99.2%

Synonym(s):

ZR006015

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zr
CAS Number:
Molecular Weight:
91.22
MDL number:
UNSPSC Code:
12141751
PubChem Substance ID:
NACRES:
NA.23

Assay

99.2%

form

powder

manufacturer/tradename

Goodfellow 858-447-40

resistivity

40 μΩ-cm, 20°C

weight

200 g

max. part. size

45 (micron)

bp

4377 °C (lit.)

mp

1852 °C (lit.)

density

6.5 g/mL at 25 °C (lit.)

SMILES string

[Zr]

InChI

1S/Zr

InChI key

QCWXUUIWCKQGHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Pyr. Sol. 1 - Water-react 1

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

M Sherif El-Eskandarany et al.
Molecules (Basel, Switzerland), 25(8) (2020-04-25)
Metallic glassy alloys with their short-range order have received considerable attention since their discovery in 1960's. The worldwide interest in metallic glassy alloys is attributed to their unique mechanical, physical, and chemical properties, which cannot be found together in long-range

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service