Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF69989023

Aluminum alloy 6082, Al97.4Si1Mg0.9Mn0.7

500mm tube, 6.35mm outside diameter, 1.63mm wall thickness, 3.09mm inside diameter, solution treated

Synonym(s):

AL227100

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352311
NACRES:
NA.23

form

tubes

manufacturer/tradename

Goodfellow 699-890-23

L × wall thickness

500 mm × 1.63 mm

O.D. × I.D.

6.35 mm × 3.09 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service