Skip to Content
MilliporeSigma

C-12644

Human Tracheal Epithelial Cells (HTEpC)

500,000 cryopreserved cells

Synonym(s):

Human Epithelial Cells

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.81
UNSPSC Code:
41106514
Biological source:
human trachea
Morphology:
(epithelial)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human trachea

packaging

pkg of 500,000 cells

morphology

(epithelial)

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Application

Primary Human Tracheal Epithelial Cells (HTEpC) are isolated from the surface epithelium of human trachea and are provided in a cryopreserved format. They stain positive for cytokeratin.The respiratory epithelia are responsible for the lubrication of the lungs, the maintenance of humidity, and the cleaning of the respiratory tract. They are an important target for drugs, toxins, and carcinogens. In addition, they are involved in many diseases such as cystic fibrosis. Consequently, HTEpC are useful for investigating the function and pathology of the respiratory system.

Biochem/physiol Actions

Trachea

Preparation Note

Click here for more information.

Analysis Note

Rigid quality control tests are performed for each lot of Epithelial Cells. They are tested for cell morphology, adherence rate and cell viability. Furthermore, flow cytometric analyses for the cell-type specific marker cytokeratin are carried out for each lot. Growth performance is tested through multiple passages under culture conditions without antibiotics and antimycotics. In addition, all cells have been tested for the absence of HIV-1, HIV-2, HBV, HCV, HTLV-1, HTLV-2 and microbial contaminants (fungi, bacteria, and mycoplasma).

Other Notes

Recommended Plating Density: 10000 - 15000 cells per cm2Passage After Thawing: P2Tested Markers: Cytokeratin positiveGuaranteed population doublings: >15
Recommended Primary Cell Culture Media:Link

Disclaimer

Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.


Still not finding the right product?


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library