Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF65793482

Lead

rod, 50mm, diameter 50mm, 99.95%

Synonym(s):

Lead, PB007960

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pb
CAS Number:
Molecular Weight:
207.20
MDL number:
UNSPSC Code:
12141723
PubChem Substance ID:
NACRES:
NA.23

Assay

99.95%

form

rod

manufacturer/tradename

Goodfellow 657-934-82

resistivity

20.65 μΩ-cm

L × diam.

50 mm × 50 mm

bp

1740 °C (lit.)

mp

327.4 °C (lit.)

SMILES string

[Pb]

InChI

1S/Pb

InChI key

WABPQHHGFIMREM-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 1A - STOT RE 1 Oral

Target Organs

Central nervous system,Blood,Immune system,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reply: Cognitive declines after deep brain stimulation are likely to be attributable to more than caudate penetration and lead location.
Karsten Witt et al.
Brain : a journal of neurology, 137(Pt 5), e275-e275 (2014-02-20)
Qinglin Chen et al.
Ecotoxicology and environmental safety, 102, 93-99 (2014-03-04)
The experiment was conducted to investigate the effects of single and combined pollution from different concentrations of atrazine (field rate, FR, 2.0 mg kg(-1) and 5 times FR, 10 mg kg(-1)) and lead (Pb) (300 mg kg(-1) and 600 mg
Paraskevi Malea et al.
Ecotoxicology and environmental safety, 104, 175-181 (2014-04-01)
The effects of lead uptake on microtubule integrity and cell viability in intermediate-juvenile leaf blades of the seagrass Cymodocea nodosa were investigated under laboratory conditions in increasing exposure concentrations (0.1, 0.25, 0.5, 5, 10, 20 and 40mg/L). Uptake kinetics was
B Ali et al.
Ecotoxicology and environmental safety, 102, 25-33 (2014-03-04)
The role of hydrogen sulfide (H2S) in alleviating lead (Pb) induced stress in oilseed rape (Brassica napus L.) was studied under laboratory conditions. Plants were grown hydroponically in greenhouse conditions under three levels (0, 100, and 400 µM) of Pb
Guillermo Horta-Puga et al.
Bulletin of environmental contamination and toxicology, 92(5), 567-573 (2014-02-11)
Lead (Pb) pollution history (1855-2001 A.D.) of the southern Gulf of Mexico (SGM) was reconstructed from the geochemical record contained in the annual bands of the hermatypic coral Orbicella faveolata from the Veracruz Reef System, Mexico. Pb concentrations ranged from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service