GF39354048
Boron
monofilament, 200m, diameter 0.1mm
Synonym(s):
Boron, B 005910
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
form
wire
manufacturer/tradename
Goodfellow 393-540-48
resistivity
1.5E12 μΩ-cm, 20 °C
L × diam.
200 m × 0.1 mm
density
2.34 g/mL at 25 °C (lit.)
SMILES string
[B]
InChI
1S/B
InChI key
ZOXJGFHDIHLPTG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Organic letters, 16(7), 1952-1955 (2014-03-29)
A novel stepwise and regioselective nucleophilic aromatic substitution (SNAr) reaction of halogenated boron dipyrrins (BODIPYs) with pyrroles has been developed under mild conditions with no catalyst needed and shown to be diversity oriented. The resultant pyrrole-substituted BODIPYs are interesting red
Chemical Society reviews, 39(11), 4388-4401 (2010-06-08)
Catalysts generated by the addition of carbon, nitrogen or phosphorus to transition metals have interesting properties and potential applications. The addition of carbon, nitrogen or phosphorus can lead to substantial modification of the catalytic efficacy of the parent metal and
Boron containing compounds as protease inhibitors.
Chemical reviews, 112(7), 4156-4220 (2012-04-24)
Plant science : an international journal of experimental plant biology, 203-204, 25-32 (2013-02-19)
Boron (B) is an essential microelement for plants and is constantly needed throughout the plant life due to its function as a structural element of the plant cell wall. B deficiency is a wide-spread problem in agricultural areas world-wide, and
Chemical Society reviews, 42(13), 5684-5713 (2013-03-30)
At the intersection of nucleic acid and boron chemistries lies a thriving world of possibilities. During the past decades, the merging of these research domains has led to fascinating discoveries in different fields ranging from material to medical sciences. In
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service