Skip to Content
MilliporeSigma
All Photos(2)

Documents

GF34237469

Erbium

rod, 100mm, diameter 6.35mm, cast, 99%

Synonym(s):

Erbium, ER007910

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Er
CAS Number:
Molecular Weight:
167.26
MDL number:
UNSPSC Code:
12141603
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

rod

manufacturer/tradename

Goodfellow 342-374-69

resistivity

86 μΩ-cm, 20°C

L × diam.

100 mm × 6.35 mm

bp

2868 °C (lit.)

mp

1529 °C (lit.)

density

9.062 g/mL at 25 °C (lit.)

SMILES string

[Er]

InChI

1S/Er

InChI key

UYAHIZSMUZPPFV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ronald J Caniglia
Facial plastic surgery clinics of North America, 12(3), 373-377 (2004-07-21)
The Erbium:YAG laser is a useful flexible tool for the surgeon interested in providing overall skin rejuvenation. With its unique wavelength, greater absorption, and less thermal injury, it can offer more precise ablation with greater safety and shorter healing times.
Isao Ishikawa et al.
Journal of periodontal research, 39(4), 275-285 (2004-06-23)
Since lasers were introduced for the treatment of oral diseases, there has been considerable advancement in technology. As a result, numerous laser systems are currently available for oral use. Neodymium:Yttrium-Aluminum:Garnet (Nd:YAG), carbon dioxide (CO(2)) laser and the semiconductor Diode lasers
Bo Li et al.
Journal of nanoscience and nanotechnology, 14(6), 4191-4195 (2014-04-18)
We have synthesized Er2S3/ZnS core/shell QDs by employing ErSt3, ZnSt2, and sulfur as precursors via a hot solution phase chemistry using a nucleation-doping strategy. X-ray diffraction (XRD), transmission electron microscope (TEM) and photoluminescence (PL) spectra were used to characterize the
G N Marcells et al.
The Journal of otolaryngology, 29(2), 78-82 (2000-05-20)
There are two lasers for resurfacing. The erbium:YAG laser and the CO2 laser both have different physical and physiologic differences when striking the skin. Therefore, each laser gives a different result. This article reviews the use of the Sharplan Silktouch
D M Clarkson
Dental update, 28(6), 298-302 (2001-08-31)
This article reviews aspects of the probable mechanisms used by erbium dental lasers for cutting dentine and enamel, describes key issues of the risk of temperature elevation and speed of cutting relative to conventional techniques and looks at issues concerned

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service