Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF17633257

Silicon-germanium alloy, Si70Ge30

50g lump, 50mm max. lump size, broken ingot

Synonym(s):

SI056100

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101711
NACRES:
NA.23

form

lumps (broken ingot)

manufacturer/tradename

Goodfellow 176-332-57

weight

50 g

max. lump size

50 mm

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service