Skip to Content
MilliporeSigma
All Photos(1)

Documents

F5062

Sigma-Aldrich

Fmoc-Lys(Me)3-OH Chloride

≥97%, for peptide synthesis

Synonym(s):

Fmoc-Lys(Me3Cl)-OH, N-alpha-Fmoc-N-alpha-trimethyl-N-L-lysine · chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H31ClN2O4
CAS Number:
Molecular Weight:
446.97
UNSPSC Code:
12352209
NACRES:
NA.26

product name

Fmoc-Lys(Me)3-OH Chloride, ≥97%

Assay

≥97%

form

liquid

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Fmoc

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C24H30N2O4.ClH/c1-26(2,3)15-9-8-14-22(23(27)28)25-24(29)30-16-21-19-12-6-4-10-17(19)18-11-5-7-13-20(18)21;/h4-7,10-13,21-22H,8-9,14-16H2,1-3H3,(H-,25,27,28,29);1H/t22-;/m0./s1

InChI key

XUJRNPVABVHOAJ-FTBISJDPSA-N

General description

Fmoc protected N-trimethyl lysine

Application

Fmoc-Lys(Me)3-OH chloride is one of the common N-terminal protected reagents used in the peptide synthesis. Some of the reported examples are:
  • Synthesis of peptide linkers for monoclonal antibody-auristatin F conjugates.
  • Preparation of multifunctional reagents with enhanced ionization properties for the analysis of protein modification in human cells and dynamic profiling of protein lipidation.
  • Sequential peptide ligation to synthesize histone H3 containing a trimethyl lysine residue, with modified tail region.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel peptide linkers for highly potent antibody? auristatin conjugate.
Doronina S O, et al.
Bioconjugate Chemistry, 19(10), 1960-1963 (2008)
Multifunctional Reagents for Quantitative Proteome?Wide Analysis of Protein Modification in Human Cells and Dynamic Profiling of Protein Lipidation During Vertebrate Development.
Broncel M, et al.
Angewandte Chemie (International Edition in English), 54(20), 5948-5951 (2015)
Sequential Peptide Ligation by Combining the Cys?Pro Ester (CPE) and Thioester Methods and Its Application to the Synthesis of Histone H3 Containing a Trimethyl Lysine Residue.
Kawakami T, et al.
Bulletin of the Chemical Society of Japan, 86(6), 690-697 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service