Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C22H21N3O4
CAS Number:
Molecular Weight:
391.42
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22
Quality Segment
product line
Novabiochem®
assay
≥98.0% (HPLC)
form
powder
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
manufacturer/tradename
Novabiochem®
application(s)
peptide synthesis
functional group
Fmoc
storage temp.
2-8°C
SMILES string
[n]1(cncc1C[C@H](NC(=O)OCC2c3c(cccc3)c4c2cccc4)C(=O)O)C
InChI
1S/C22H21N3O4/c1-25-13-23-11-14(25)10-20(21(26)27)24-22(28)29-12-19-17-8-4-2-6-15(17)16-7-3-5-9-18(16)19/h2-9,11,13,19-20H,10,12H2,1H3,(H,24,28)(H,26,27)/t20-/m0/s1
InChI key
UEDYXEZHNPXCFB-FQEVSTJZSA-N
General description
Building block for the incorporation of 1-methylhistidine by Fmoc SPPS.
Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Analysis Note
Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Assay (HPLC, area%): ≥ 98.0 % (a/a)
Solubility (0,5 mmol in 4 ml NMP + 500 µl DIPEA): clearly soluble
Appearance of substance (visual): powder
Identity (IR): passes test
Assay (HPLC, area%): ≥ 98.0 % (a/a)
Solubility (0,5 mmol in 4 ml NMP + 500 µl DIPEA): clearly soluble
Legal Information
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.