Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F4438

Sigma-Aldrich

Fmoc-Lys(Me,Boc)-OH

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H34N2O6
Molecular Weight:
482.57
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Quality Level

Assay

≥97%

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Boc
Fmoc

shipped in

dry ice

storage temp.

−20°C

SMILES string

CN(CCCC[C@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O)C(=O)OC(C)(C)C

InChI

1S/C27H34N2O6/c1-27(2,3)35-26(33)29(4)16-10-9-15-23(24(30)31)28-25(32)34-17-22-20-13-7-5-11-18(20)19-12-6-8-14-21(19)22/h5-8,11-14,22-23H,9-10,15-17H2,1-4H3,(H,28,32)(H,30,31)/t23-/m0/s1

InChI key

JHMSFOFHTAYQLS-QHCPKHFHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fmoc protected N-methylated lysine

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service