Skip to Content
MilliporeSigma
All Photos(2)

Documents

F15331

Sigma-Aldrich

4-Fluorotoluene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4F
CAS Number:
Molecular Weight:
110.13
Beilstein:
1362373
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.468 (lit.)

bp

116 °C (lit.)

mp

−56 °C (lit.)

density

1 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(F)cc1

InChI

1S/C7H7F/c1-6-2-4-7(8)5-3-6/h2-5H,1H3

InChI key

WRWPPGUCZBJXKX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William D Tuttle et al.
The Journal of chemical physics, 146(24), 244310-244310 (2017-07-03)
A study of the vibration and vibration-torsion levels of para-fluorotoluene in the 580-830 cm
Isaiah R Speight et al.
Molecules (Basel, Switzerland), 25(3) (2020-02-06)
Owing to the strength of the C-F bond, the 'direct' preparation of Grignard reagents, i.e., the interaction of elemental magnesium with an organic halide, typically in an ethereal solvent, fails for bulk magnesium and organofluorine compounds. Previously described mechanochemical methods

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service