Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

547948

Sigma-Aldrich

α,α,α-Trifluorotoluene

anhydrous, ≥99%

Synonym(s):

Benzotrifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CF3
CAS Number:
Molecular Weight:
146.11
Beilstein:
1906908
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
technique(s):
NMR: suitable
bp:
102 °C (lit.)
vapor pressure:
38.83 mmHg

grade

anhydrous

Quality Level

vapor density

5.04 (vs air)

vapor pressure

38.83 mmHg

Assay

≥99%

form

liquid

expl. lim.

0.34-6.3 % (lit.)

technique(s)

NMR: suitable

impurities

≤0.001% water
water

refractive index

n20/D 1.414 (lit.)

bp

102 °C (lit.)

mp

−29 °C (lit.)

density

1.19 g/mL at 20 °C (lit.)

SMILES string

FC(F)(F)c1ccccc1

InChI

1S/C7H5F3/c8-7(9,10)6-4-2-1-3-5-6/h1-5H

InChI key

GETTZEONDQJALK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α,α,α-Trifluorotoluene (TFT) is a potential substitute solvent for reactions carried out in dichloromethane and related solvents. It is also applicable in fluorous synthesis as an organic/fluorous hybrid solvent. TFT undergoes photocycloaddition with cyclopentene to form cyano-substituted tetracyclo[6.3.0.02,11.03,7]undec-9-enes. The nuclear magnetic resonance spectral data of TFT has been reported. The bimolecular rate constants of the aqueous phase reactions between sulfate and phosphate radicals with TFT have been determined by conventional flash-photolysis.

Application

α,α,α-Trifluorotoluene may be used as an internal standard for 19F NMR spectroscopic determination of copper-catalyzed trifluoromethylation of alkenes.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Liq. 2 - STOT RE 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

46.4 °F - closed cup

Flash Point(C)

8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

α,α,α-Trifluorotoluene.
Ogawa A and Tsuchii K. <BR/>
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2006)
Reaction of sulfate and phosphate radicals with α,α,α-trifluorotoluene.
Rosso JA, et al.
J. Chem. Soc. Perkin Trans. II, 2, 205-210 (1999)
The meta photocycloaddition of benzonitrile and a,a,a-trifluorotoluene to cyclopentene.
Osselton EM and Cornelisse J.
Tetrahedron Letters, 26(4), 527-530 (1985)
Copper-Catalyzed Trifluoromethylation-Initiated Radical 1,2-Aryl Migration in α,α-Diaryl Allylic Alcohols.
Liu X, et al.
Angewandte Chemie (International Edition in English), 125(27), 7100-7104 (2013)
Nuclear magnetic resonance spectral parameters of α,α,α-trifluorotoluene.
Kostelnik RJ, et al.
Canadian Journal of Chemistry, 47(18), 3313-3318 (1969)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service