Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

DWKBGE814PNC

Azlon Dropper Bottles with spouted closure and cap

round bottom, low-density polyethylene, capacity 30 mL

Synonym(s):

BGE814PNC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121819
NACRES:
NB.43

material

low-density polyethylene
round bottom

packaging

pkg of 10 ea

manufacturer/tradename

Azlon

capacity

30 mL

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for as dropping bottle and excellent for dispensing gels

Features and Benefits

Pivotting nozzle closure

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service