Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DWK374324

Wheaton® Polypropylene Nozzle & Glass Sinker

Synonym(s):

374324

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104812
NACRES:
NB.43

material

polypropylene nozzle (2 and 3 mm)

Sterilization Compatibility

autoclavable compatible

packaging

pkg of 6 ea

manufacturer/tradename

WHEATON® 374324

Looking for similar products? Visit Product Comparison Guide

Application

Research, Development and Quality Control, Pharmaceutical and Biotechnology, Food and Beverage, Waste Water and Sample Collection

Features and Benefits

  • The nozzle connects to the pump tubing for dispensing of liquids
  • The glass sinker is used to secure tubing when drawing from a container

Legal Information

WHEATON is a registered trademark of Wheaton Industries, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service