Key Documents
DWK11-6200-S
Wheaton® Microliter Vial
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
material
V-bottom polypropylene bottom
amber borosilicate glass Type 1 vial (Silanized, with Marking Patch)
packaging
pkg of 100 ea
manufacturer/tradename
WHEATON® 11-6200-S
D × H
12 mm × 32 mm
cap size
11 mm
volume
2.0 mL
Application
Chromatography
Features and Benefits
  • Double ring finish for added closure performance.
  • Approved for LC & GC.
  • Where noted, Certified Class 10,000 clean room packed enhances data integrity.
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Related Content
Explore our protocol for suspension primary hepatocyte cell culture. Discover our primary human hepatocytes and plating and maintenance media formulations.
Explore our protocol for suspension primary hepatocyte cell culture. Discover our primary human hepatocytes and plating and maintenance media formulations.
Explore our protocol for suspension primary hepatocyte cell culture. Discover our primary human hepatocytes and plating and maintenance media formulations.
Explore our protocol for suspension primary hepatocyte cell culture. Discover our primary human hepatocytes and plating and maintenance media formulations.