Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

D93600

Sigma-Aldrich

O,O′-Diethyl dithiophosphate

technical grade, 90%

Synonym(s):

Di-O-ethyl dithiophosphate, Dithiophosphoric acid O,O-diethyl ester, O,O-Diethyl dithiophosphate, O,O-Diethyl dithiophosphoric acid ester, O,O-Diethyl hydrogen phosphorodithioate, O,O-Diethyl phosphorodithioate, O,O-Diethyl thiolothionophosphate, O,O′-Diethyl hydrogen dithiophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)2P(S)SH
CAS Number:
Molecular Weight:
186.23
Beilstein:
507407
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

form

liquid

refractive index

n20/D 1.512 (lit.)

bp

60 °C/1 mmHg (lit.)

density

1.11 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOP(S)(=S)OCC

InChI

1S/C4H11O2PS2/c1-3-5-7(8,9)6-4-2/h3-4H2,1-2H3,(H,8,9)

InChI key

IRDLUHRVLVEUHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-Shing Huang et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 22(10), 1375-1378 (2006-10-14)
Inductively coupled plasma-mass spectrometry (ICP-MS) incorporated with an on-line preconcentration system was used to determine trace amounts of ten metals including Ni, Cu, Zn, Rh, Ag, Cd, In, Au, Tl, and Pb in aqueous solutions and seawater. These metals, which
César Ricardo Teixeira Tarley et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 21(11), 1337-1342 (2005-12-02)
The present paper describes the on-line coupling of a flow-injection system to a new technique, thermospray flame furnace-AAS (TS-FF-AAS), for the preconcentration and determination of copper in water samples. Copper was preconcentrated onto polyurethane foam (PUF) complexed with ammonium O,O-diethyldithiophosphate
Jeferson Schneider Carletto et al.
Analytica chimica acta, 638(1), 45-50 (2009-03-21)
A hollow fiber renewal liquid membrane (HFRLM) extraction method to determine cadmium (II) in water samples using Flame Atomic Absorption Spectrometry (FAAS) was developed. Ammonium O,O-diethyl dithiophosphate (DDTP) was used to complex cadmium (II) in an acid medium to obtain
K Borg et al.
Archives of toxicology, 62(5), 387-391 (1988-01-01)
Brown trout, Salmo trutta, were exposed to water containing 0.1 microgram/l 203Hg2+, alone or with potassium ethylxanthate (PEX), sodium isopropylxanthate (SIX), sodium diethyldithiophosphate (SEP), sodium diisopropyldithiophosphate (SIP), sodium dimethyldithiocarbamate (SMC), sodium diethyldithiocarbamate (SEC) or sodium pyridinethione (SPyr), respectively. After 1
Neeranuch Suwannarin et al.
International journal of environmental research and public health, 17(21) (2020-10-31)
Exposure to insecticides containing organophosphate (OP) and neonicotinoid (NEO) compounds has been associated with adverse reproductive health outcomes. This study characterized and identified predictors of exposure to OP and NEO among 100 reproductive-age farmworkers from two intensive farming areas in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service