Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CDS014665

Sigma-Aldrich

dimethyl phosphate

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H7O4P
Molecular Weight:
126.05
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

liquid

SMILES string

O=P(OC)(OC)O

InChI

1S/C2H7O4P/c1-5-7(3,4)6-2/h1-2H3,(H,3,4)

InChI key

KKUKTXOBAWVSHC-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abigail Gaylord et al.
Molecular and cellular endocrinology, 502, 110666-110666 (2020-01-19)
Endocrine disrupting chemicals are known to cause neurodevelopmental toxicity through direct and indirect pathways. In this study we used data from the National Health and Nutrition Examination Surveys, along with known exposure-disease relationships, to quantify the intellectual disability burden attributable
Yan Zhang et al.
American journal of epidemiology, 189(5), 375-383 (2019-12-18)
Organophosphates (OPs) are the most heavily used pesticides in China. The Chinese population, including preconceptional women, is highly exposed, yet little is known regarding the associations between OP exposure and menstruation in humans. We conducted a cross-sectional analysis in women
Yannick Philipp Stenzel et al.
Journal of chromatography. A, 1592, 188-191 (2019-02-18)
Nowadays, lithium ion batteries (LIBs) are the preferred energy supply for consumer electronics and electric vehicles. But, intrinsic reactions in the LIB system lead to a reduced battery life and impaired safety properties. Organo(fluoro)phosphates (O(F)Ps) as decomposition product of the
Ian E Cock et al.
Pharmacognosy research, 3(2), 85-94 (2011-07-21)
Scaevola spinescens is an endemic Australian native plant with a history of use as a medicinal agent by indigenous Australians. Yet the medicinal bioactivities of this plant are poorly studied. S. spinescens solvent extracts were tested for antimicrobial activity, antiviral
Taylor M Etzel et al.
Environment international, 142, 105858-105858 (2020-07-01)
Animal studies suggest that organophosphorus pesticides (OPs) may be environmental obesogens. While prenatal OP exposures have been associated with altered infant glucose metabolism, associations with pediatric adiposity remain unknown. We summed concentrations of three dimethylphosphate (∑DMP) and three diethylphosphate (∑DEP)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service