Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D51401

Sigma-Aldrich

Diethyl 1,2-hydrazinedicarboxylate

97%

Synonym(s):

1,2-Dicarbethoxyhydrazine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCONHNHCOOC2H5
CAS Number:
Molecular Weight:
176.17
Beilstein:
1781585
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

bp

250 °C (lit.)

mp

131-133 °C (lit.)

SMILES string

CCOC(=O)NNC(=O)OCC

InChI

1S/C6H12N2O4/c1-3-11-5(9)7-8-6(10)12-4-2/h3-4H2,1-2H3,(H,7,9)(H,8,10)

InChI key

JXMLAPZRDDWRRV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service