Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E16503

Sigma-Aldrich

Ethyl carbazate

97%

Synonym(s):

Hydrazinocarboxylic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NNHCO2C2H5
CAS Number:
Molecular Weight:
104.11
Beilstein:
878265
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

bp

108-110 °C/22 mmHg (lit.)

mp

44-47 °C (lit.)

SMILES string

CCOC(=O)NN

InChI

1S/C3H8N2O2/c1-2-7-3(6)5-4/h2,4H2,1H3,(H,5,6)

InChI key

VYSYZMNJHYOXGN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Felix Sellberg et al.
PloS one, 14(12), e0225777-e0225777 (2019-12-07)
The objective of this study was to investigate whether a soluble polymer and aldehyde-scavenger, polyvinylalcohol-carbazate (PVAC), can inhibit hemolysis in the storage of red blood cells (RBC). The effect of PVAC was assessed over a wide range of concentrations, using
Yun Seop Kim et al.
Biomaterials science, 6(5), 1020-1030 (2018-04-05)
The proteolytic microenvironment in the wound area reduces the stability and the half-life of growth factors in vivo, making difficult the topical delivery of growth factors. Here, epidermal growth factor (EGF) was conjugated to hyaluronate (HA) to improve the long-term
Hyunkyu Kim et al.
Carbohydrate polymers, 252, 117132-117132 (2020-11-14)
Transdermal immunomodulation is of increasing interest as an efficient drug delivery method. It non-invasively delivers drugs directly to skin-resident immune cells, thereby avoiding the first-pass metabolism. Herein, we prepared ovalbumin-conjugated hyaluronic acid-methacrylate (HAMA-OVA) and schizophyllan-methacrylate (SPGMA) hybrid nanogels and investigated
Yun Seop Kim et al.
Journal of controlled release : official journal of the Controlled Release Society, 307, 76-89 (2019-06-24)
The intranasal drug administration has attracted great interest as a non-invasive route allowing targeted delivery of drugs directly to the brain. However, one of the main issues in nasal drug administration is mucociliary clearance. Hyaluronate (HA) has been widely used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service